Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

374342

Sigma-Aldrich

N,N′,N′′-Tritosyldiethylenetriamine disodium salt

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3C6H4SO2N(Na)CH2CH2]2NSO2C6H4CH3
CAS Number:
Molecular Weight:
609.69
Beilstein/REAXYS Number:
3585044
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

275 °C (dec.) (lit.)

solubility

95% ethanol: soluble 25 mg/mL, clear, colorless to yellow

functional group

amine
sulfonamide

SMILES string

Cc1ccc(cc1)S(=O)(=O)N([Na])CCN(CCN([Na])S(=O)(=O)c2ccc(C)cc2)S(=O)(=O)c3ccc(C)cc3

InChI

1S/C25H29N3O6S3.2Na/c1-20-4-10-23(11-5-20)35(29,30)26-16-18-28(37(33,34)25-14-8-22(3)9-15-25)19-17-27-36(31,32)24-12-6-21(2)7-13-24;;/h4-15H,16-19H2,1-3H3;;/q-2;2*+1

InChI key

HWXFIPYUNYRUJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′,N′′-Tritosyldiethylenetriamine is a tosyl derivative and its complexation with Ag(I) and Cu(II) is reported.[1]

Application

N,N′,N′′-Tritosyldiethylenetriamine disodium salt may be used in the preparation of tritosylate of 1,4,7-triazacyclononane, via reaction with 1,2-dibromoethane in DMF.[2] It may be used in the preparation of 1,4,7-triazacyclononane, via reaction with ditosyl derivative of 1,2-ethanediol.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability constants of some macrocyclic complexes of Ag (I) and Cu (II) in mixed solvents by potentiometry.
Sil A, et al.
Supramolecular Chemistry, 15(6), 451-457 (2003)
Thorsten Allscher et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 8), o1734-o1734 (2009-01-01)
The title compound, C(6)H(16)N(3) (+)·Br(-), is the bromide of the monoprotonated aza-macrocyclic triamine 1,4,7-triaza-cyclo-nonane (tacn). The threefold axis of the triamine is broken by the protonation of one of the three amine functions. The ammonium proton is bonded in an
Acid dissociation kinetics of copper (II) complexes of 1, 4, 7-triazacyclononane ([9] aneN3).
Hay RW and Pujari MP.
Transition Met. Chem. (London), 17(2), 81-83 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service