368520
Tris(2-benzimidazolylmethyl)amine
technical grade, 92%
Synonym(s):
(BimH)3, N,N-Bis(1H-benzimidazol-2-ylmethyl)-1H-benzimidazole-2-methanamine
About This Item
grade
technical grade
assay
92%
mp
277-281 °C (lit.)
SMILES string
C(N(Cc1nc2ccccc2[nH]1)Cc3nc4ccccc4[nH]3)c5nc6ccccc6[nH]5
InChI
1S/C24H21N7/c1-2-8-17-16(7-1)25-22(26-17)13-31(14-23-27-18-9-3-4-10-19(18)28-23)15-24-29-20-11-5-6-12-21(20)30-24/h1-12H,13-15H2,(H,25,26)(H,27,28)(H,29,30)
InChI key
YQIGEJHOYBUSLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Carbonic anhydrase models
- Porous lanthanide tris(benzimidazolylmethyl)amine nitrato frameworks
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service