Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

365254

Supelco

Potassium tetrakis(4-chlorophenyl)borate

98%

Synonym(s):

Tetrakis(4-chlorophenyl)boron potassium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)4BK
CAS Number:
Molecular Weight:
496.11
Beilstein/REAXYS Number:
4117827
EC Number:
MDL number:
UNSPSC Code:
26111700

assay

98%

SMILES string

[K+].Clc1ccc(cc1)[B-](c2ccc(Cl)cc2)(c3ccc(Cl)cc3)c4ccc(Cl)cc4

InChI

1S/C24H16BCl4.K/c26-21-9-1-17(2-10-21)25(18-3-11-22(27)12-4-18,19-5-13-23(28)14-6-19)20-7-15-24(29)16-8-20;/h1-16H;/q-1;+1

InChI key

SAGICZRAKJSWLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service