Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

363499

Sigma-Aldrich

1-Phenyl-1-hexyne

99%

Synonym(s):

1-Hexynylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡C(CH2)3CH3
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

refractive index

n20/D 1.5349 (lit.)

bp

229-232 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CCCCC#Cc1ccccc1

InChI

1S/C12H14/c1-2-3-4-6-9-12-10-7-5-8-11-12/h5,7-8,10-11H,2-4H2,1H3

InChI key

VBRLZTLFLNZEPZ-UHFFFAOYSA-N

General description

1-Phenyl-1-hexyne is an alkyne. Stereoselective hydrogenation of 1-phenyl-1-hexyne at 298K and atmospheric pressure of H2 over Pd catalysts supported on mesostructured silica is reported.[1] Polymerization of 1-phenyl-1-hexyne by pentahalides of niobium and tanatalum has been reported.[2] Electrolytically induced, base-catalyzed isomerization of 1-phenyl-1-hexyne to 1-phenyl-1, 2-hexadiene in dimethylformamide containing tetra-n-butylammonium perchlorate using glassy carbon rotating ring-disk electrode has been reported.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization of 1-phenyl-1-alkynes by halides of niobium and tantalum.
Masuda T, et al.
Macromolecules, 18(3), 311-317 (1985)
Mesostructured silicas as supports for palladium-catalyzed hydrogenation of phenyl acetylene and 1-phenyl-1-hexyne to alkenes.
Marin-Astorga N, et al.
J. Mol. Catal. A: Chem., 247(1), 145-152 (2006)
Characterization of the electrolytically induced isomerization of 1-phenyl-1-hexyne: Part II. Hydrodynamic voltammetry with rotating disk and ring-disk electrodes.
Stemple JZ and Peters DG.
J. Electroanal. Chem. Interfac. Electrochem., 286(1), 109-121 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service