360007
2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine
98%
Synonym(s):
BEMP
About This Item
assay
98%
refractive index
n20/D 1.477 (lit.)
bp
74 °C/0.03 mmHg (lit.)
density
0.948 g/mL at 25 °C (lit.)
SMILES string
CCN(CC)P1(=NC(C)(C)C)N(C)CCCN1C
InChI
1S/C13H31N4P/c1-8-17(9-2)18(14-13(3,4)5)15(6)11-10-12-16(18)7/h8-12H2,1-7H3
InChI key
VSCBATMPTLKTOV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service