Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

347027

Sigma-Aldrich

4-Amino-3-methoxybenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(OCH3)CO2H
CAS Number:
Molecular Weight:
167.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

186-188 °C (lit.)

application(s)

peptide synthesis

SMILES string

COc1cc(ccc1N)C(O)=O

InChI

1S/C8H9NO3/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4H,9H2,1H3,(H,10,11)

InChI key

JNFGLYJROFAOQP-UHFFFAOYSA-N

Application

Key building block for the efficient synthesis of carbazole alkaloids mukonine and mukonidine via oxidative annulation.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domenico Pirone et al.
Polymers, 12(5) (2020-05-06)
A molecular design approach was used to create asymmetrical visible light-triggered azo-derivatives that can be good candidates for polymer functionalization. The specific electron-donor substituted molecules were characterized and studied by means of NMR analyses and UV-visible spectroscopy, comparing the results
Tetrahedron, 59, 5317-5317 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service