Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346497

Sigma-Aldrich

2-Fluoro-3-(trifluoromethyl)benzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)CHO
CAS Number:
Molecular Weight:
192.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.455 (lit.)

density

1.393 g/mL at 25 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

Fc1c(C=O)cccc1C(F)(F)F

InChI

1S/C8H4F4O/c9-7-5(4-13)2-1-3-6(7)8(10,11)12/h1-4H

InChI key

XDMZVNQKVMTCSP-UHFFFAOYSA-N

Application

2-Fluoro-3-(trifluoromethyl)benzaldehyde was used in the synthesis of three series of curcumin analogs with mono-carbonyl.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang Liang et al.
Chemical & pharmaceutical bulletin, 56(2), 162-167 (2008-02-02)
The synthesis of three series of curcumin analogues with mono-carbonyl is described. Their in vitro anti-bacterial activities against seven Gram-positive and Gram-negative bacteria were tested and the effect of substituents on the aryl ring and the space structure of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service