Skip to Content
Merck
All Photos(1)

Key Documents

346195

Sigma-Aldrich

Dibenzofuran

technical grade, 90%

Synonym(s):

Diphenylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O
CAS Number:
Molecular Weight:
168.19
Beilstein/REAXYS Number:
121100
EC Number:
MDL number:
UNSPSC Code:
12352100

Pricing and availability is not currently available.

grade

technical grade

assay

90%

bp

154-155 °C/20 mmHg (lit.)

mp

80-82 °C (lit.)

SMILES string

c1ccc2c(c1)oc3ccccc23

InChI

1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

TXCDCPKCNAJMEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service