Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

340049

Sigma-Aldrich

DIPSO

97%

Synonym(s):

3-(N,N-Bis[2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid, N,N-Bis(2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
Beilstein/REAXYS Number:
2105454
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

assay

97%

useful pH range

7.0-8.2

pKa (25 °C)

7.6

mp

189-192 °C (lit.)

SMILES string

OCCN(CCO)CC(O)CS(O)(=O)=O

InChI

1S/C7H17NO6S/c9-3-1-8(2-4-10)5-7(11)6-15(12,13)14/h7,9-11H,1-6H2,(H,12,13,14)

InChI key

XCBLFURAFHFFJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service