Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338699

Sigma-Aldrich

Dipentylamine, mixture of isomers

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)4]2NH
CAS Number:
Molecular Weight:
157.30
Beilstein/REAXYS Number:
906746
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.42 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)

assay

99%

refractive index

n20/D 1.4258 (lit.)

bp

202-203 °C (lit.)

density

0.767 g/mL at 25 °C

SMILES string

CCCCCNCCCCC

InChI

1S/C10H23N/c1-3-5-7-9-11-10-8-6-4-2/h11H,3-10H2,1-2H3

InChI key

JACMPVXHEARCBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dipentylamine was used in the synthesis of new melt-spinnable polymeric precursor to boron nitride ceramic fibers. It was used to compose background electrolyte for the separation of linear alkylbenzene sulfonates by nonaqueous capillary electrophoresis. It was employed as organic additive in the synthesis of pure AlP04-H2 (aluminophosphate material).

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

156.2 °F - closed cup

flash_point_c

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Herrero-Martínez et al.
Electrophoresis, 22(10), 2017-2024 (2001-07-24)
The separation of linear alkylbenzene sulfonates (LAS) by nonaqueous capillary electrophoresis (NACE) using negative polarity, and a buffer containing acetic acid and an alkylamine in nonaqueous ethanol, has been investigated. Several primary, secondary, and tertiary alkylamines with alkyl chains of
Dipentylamine-modified polyborazylene: a new, melt-spinnable polymeric precursor to boron nitride ceramic fibers.
Wideman T and Sneddon LG.
Chemistry of Materials, 8(1), 3-5 (1996)
Further studies on aluminophosphate molecular sieves. Part 2.-VPI-5 and related aluminophosphate materials.
Li H-X and Davis ME.
J. Chem. Soc., Faraday, 89(6), 957-964 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service