Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330418

Sigma-Aldrich

N-Phthaloyl-L-glutamic acid

99% (HPLC)

Synonym(s):

PhGA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO6
CAS Number:
Molecular Weight:
277.23
Beilstein/REAXYS Number:
90136
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

99% (HPLC)

form

powder or crystals

optical activity

[α]25/D -45 to -41°, c = 10 g/mL in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

160-162 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CC[C@H](N1C(=O)c2ccccc2C1=O)C(O)=O

InChI

1S/C13H11NO6/c15-10(16)6-5-9(13(19)20)14-11(17)7-3-1-2-4-8(7)12(14)18/h1-4,9H,5-6H2,(H,15,16)(H,19,20)/t9-/m0/s1

InChI key

FEFFSKLJNYRHQN-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service