Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329401

Sigma-Aldrich

5-Chloro-2′-deoxyuridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11ClN2O5
CAS Number:
Molecular Weight:
262.65
MDL number:

assay

98%

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Cl)C(=O)NC2=O

InChI

1S/C9H11ClN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1

InChI key

NJCXGFKPQSFZIB-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian Tian et al.
Molecular cell, 81(1), 198-211 (2020-12-10)
Replication fork reversal is a global response to replication stress in mammalian cells, but precisely how it occurs remains poorly understood. Here, we show that, upon replication stress, DNA topoisomerase IIalpha (TOP2A) is recruited to stalled forks in a manner
Shan Li et al.
Molecular cell, 74(6), 1123-1137 (2019-05-06)
Abnormal processing of stressed replication forks by nucleases can cause fork collapse, genomic instability, and cell death. Despite its importance, it is poorly understood how the cell properly controls nucleases to prevent detrimental fork processing. Here, we report a signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service