Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

329126

Sigma-Aldrich

4-Methyl-1,2,4-triazoline-3,5-dione

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3N3O2
CAS Number:
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

mp

107-109 °C (lit.)

SMILES string

CN1C(=O)N=NC1=O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangbo Kong et al.
Se pu = Chinese journal of chromatography, 22(2), 97-100 (2005-02-17)
The dienophile 4-methyl-1,2,4-triazoline-3,5-dione (MTAD) adducts of conjugated diene compounds were used to locate the position of the conjugated double bonds in the parent compounds by high-resolution gas chromatography and electron impact mass spectrometry. Their mass spectra are marked by recognizable
D C Young et al.
Journal of chromatography, 522, 295-302 (1990-11-28)
The dienophile 4-methyl-1,2,4-triazoline-3,5-dione forms stable adducts with conjugated dienes by generating Diels-Alder cycloaddition products. The reaction is rapid, highly selective for conjugated dienes and the derivatives are suitable for analysis by gas chromatography. Their mass spectra are marked by their
J L Sébédio et al.
Biochimica et biophysica acta, 1345(1), 5-10 (1997-03-10)
Metabolism of conjugated isomers of linoleic acid (CLA) in rats was studied by feeding high quantities of CLA (180 mg/day) for six days to animals that had been reared on a fat-free diet for two weeks. After this period, animals
Phil S Baran et al.
Organic letters, 5(11), 1999-2001 (2003-05-24)
[reaction: see text] The scope and generality of a new reaction of indoles with MTAD is discussed. In most cases the ene-type reaction proceeds within seconds or minutes at 0 degrees C to provide the urazole adducts in high yield.
B Yeung et al.
Journal of chromatography, 645(1), 115-123 (1993-08-13)
A mass spectrometric method for the detection of vitamin D3 metabolites is described. This method involves the derivatization of the metabolites by cycloaddition with 4-phenyl-1,2,4-triazoline-3,5-dione, followed by their characterization by continuous-flow fast atom bombardment (CF-FAB) tandem mass spectrometry (MS-MS) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service