Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328723

Sigma-Aldrich

4-Fluorophenyl chloroformate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2C6H4F
CAS Number:
Molecular Weight:
174.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.4933 (lit.)

density

1.329 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccc(OC(Cl)=O)cc1

InChI

1S/C7H4ClFO2/c8-7(10)11-6-3-1-5(9)2-4-6/h1-4H

InChI key

MSBGPEACXKBQSX-UHFFFAOYSA-N

Application

4-Fluorophenyl chloroformate may be used in chemical synthesis.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

168.8 °F - closed cup

flash_point_c

76 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service