Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

323535

Sigma-Aldrich

Ammonium hexachloropalladate(IV)

99.95% trace metals basis

Synonym(s):

Palladium(IV)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2PdCl6
CAS Number:
Molecular Weight:
355.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.95% trace metals basis
form:
solid

assay

99.95% trace metals basis

form

solid

impurities

≤550.0 ppm Trace Metal Analysis

SMILES string

[NH4+].[NH4+].Cl[Pd--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2H3N.Pd/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

LOLIPEAFAJNGJM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J N Hooper et al.
Physical chemistry chemical physics : PCCP, 20(41), 26734-26743 (2018-10-17)
The ability to clearly relate local structure to function is desirable for many catalytically relevant Pd-containing systems. This report represents the first direct 105Pd solid state NMR measurements of diamagnetic inorganic (K2Pd(iv)Cl6, (NH4)2Pd(iv)Cl6 and K2Pd(iv)Br6) complexes, and micron- and nano-sized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service