Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

306371

Sigma-Aldrich

2-(Chloromethyl)tetrahydro-2H-pyran

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClO
CAS Number:
Molecular Weight:
134.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

refractive index

n20/D 1.462 (lit.)

bp

53-54 °C/12 mmHg (lit.)

density

1.075 g/mL at 25 °C (lit.)

functional group

chloro
ether

SMILES string

ClCC1CCCCO1

InChI

1S/C6H11ClO/c7-5-6-3-1-2-4-8-6/h6H,1-5H2

InChI key

PPYKTTGONDVGPX-UHFFFAOYSA-N

Application

2-(Chloromethyl)tetrahydro-2H-pyran may be used in chemical synthesis studies.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhao et al.
Cell metabolism, 31(5), 937-955 (2020-04-24)
Cell proliferation and inflammation are two metabolically demanding biological processes. How these competing processes are selectively executed in the same cell remains unknown. Here, we report that the enzyme carbamoyl-phosphate synthetase, aspartyl transcarbamoylase, and dihydroorotase (CAD) deamidates the RelA subunit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service