Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

305073

Sigma-Aldrich

4-Heptylaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6C6H4NH2
CAS Number:
Molecular Weight:
191.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

159 °C/18 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCc1ccc(N)cc1

InChI

1S/C13H21N/c1-2-3-4-5-6-7-12-8-10-13(14)11-9-12/h8-11H,2-7,14H2,1H3

InChI key

BNEWZYZRLNNWNR-UHFFFAOYSA-N

General description

Adsorption of 4-heptylaniline on dodecyl-grafted mesoporous alumina (C12-MPA) was studied.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular selective adsorption of alkylphenols and alkylanilines from water by alkyl-grafted mesoporous alumina: A comparative study to alkyl-grafted mesoporous silica.
Inumaru K, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 95(1), 279-285 (2006)
Kristiana T Santoso et al.
Bioorganic & medicinal chemistry, 27(16), 3532-3545 (2019-07-03)
A library of thirty-two quinolinequinones (QQs) with various amine substituents at the 6- and 7-positions were synthesised efficiently and in good yields for evaluation as potential anti-tuberculosis agents. Mycobacterium tuberculosis growth inhibition assays demonstrated that QQs bearing moderate length alkyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service