Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

301922

Sigma-Aldrich

2-Acetamido-4-methylthiazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2OS
CAS Number:
Molecular Weight:
156.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

98%

form

solid

mp

134-136 °C (lit.)

functional group

amide

SMILES string

CC(=O)Nc1nc(C)cs1

InChI

1S/C6H8N2OS/c1-4-3-10-6(7-4)8-5(2)9/h3H,1-2H3,(H,7,8,9)

Inchi Key

DPDJXTANWGNJOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The metabolism of 2-acetamido-4-methylthiazole in rat has been studied.[1] Sulfonation of 2-acetamido-4-methylthiazole by chlorosulfonic acid has been reported.[2]

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Chatfield et al.
The Biochemical journal, 134(4), 869-878 (1973-08-01)
The metabolism of some anti-inflammatory acetamidothiazoles was studied in the rat. The main metabolites were the corresponding acetylthiohydantoic acids, produced by fission of the thiazole ring. Minor metabolites arising from oxidation of the methyl or phenyl substituents were also identified.
Notes-2-Aminothiazolesulfonamides.
Ziegler C, et al.
The Journal of Organic Chemistry, 25(8), 1454-1455 (1960)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service