Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300470

Sigma-Aldrich

(2R,3R)-trans-3-Phenyloxirane-2-methanol

97%

Synonym(s):

(2R,3R)-(+)-3-Phenylglycidol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
5253267
MDL number:
UNSPSC Code:
12352103

assay

97%

optical activity

[α]20/D +49°, c = 2 in chloroform

mp

51-53 °C (lit.)

SMILES string

OC[C@H]1O[C@@H]1c2ccccc2

Looking for similar products? Visit Product Comparison Guide

Legal Information

Licensed under U.S. Patent 4 471 130

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service