Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

296341

Sigma-Aldrich

1,2:3,5-Di-O-isopropylidene-D-xylofuranose

99%

Synonym(s):

D-Xylose diacetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O5
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

SMILES string

CC1(C)OCC2OC3OC(C)(C)OC3C2O1

InChI

1S/C11H18O5/c1-10(2)12-5-6-7(14-10)8-9(13-6)16-11(3,4)15-8/h6-9H,5H2,1-4H3

InChI key

NKZDPBSWYPINNF-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service