Skip to Content
MilliporeSigma
All Photos(1)

Documents

295078

Sigma-Aldrich

cis-2-Butene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH3
CAS Number:
Molecular Weight:
56.11
Beilstein/REAXYS Number:
1361341
EC Number:
MDL number:
UNSPSC Code:
12142100

vapor density

2 (vs air)

vapor pressure

1414 mmHg ( 21 °C)

assay

95%

autoignition temp.

615 °F

expl. lim.

9.7 %

bp

3.7 °C (lit.)

mp

−139 °C (lit.)

SMILES string

[H]\C(C)=C(/[H])C

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3-

InChI key

IAQRGUVFOMOMEM-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Brass regulator Z146706 or Z146714 is recommended.

replaced by

Product No.
Description
Pricing

pictograms

FlameGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service