Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

291579

Sigma-Aldrich

Lithium tetraphenylborate tris(1,2-dimethoxyethane)

98%

Synonym(s):

Tetraphenylboron lithium tris(1,2-dimethoxyethane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4BLi·3CH3O(CH2CH2)OCH3
CAS Number:
Molecular Weight:
596.53
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

reaction suitability

core: boron
reagent type: catalyst

SMILES string

[Li+].COCCOC.COCCOC.COCCOC.c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.3C4H10O2.Li/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;3*1-5-3-4-6-2;/h1-20H;3*3-4H2,1-2H3;/q-1;;;;+1

InChI key

ADYUZFWVPWDPFK-UHFFFAOYSA-N

application

  • Reactant for preparation of dinuclear lithium aqua DMSO complex

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service