Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

291536

Sigma-Aldrich

Bis(2,2,2-trichloroethyl) azodicarboxylate

≥97%

Synonym(s):

Azodicarboxylic acid bis(2,2,2-trichloroethyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl3CH2OCON=NCOOCH2CCl3
CAS Number:
Molecular Weight:
380.82
Beilstein/REAXYS Number:
2141527
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97%

form

solid

mp

109-111 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)COC(=O)\N=N\C(=O)OCC(Cl)(Cl)Cl

InChI

1S/C6H4Cl6N2O4/c7-5(8,9)1-17-3(15)13-14-4(16)18-2-6(10,11)12/h1-2H2/b14-13+

InChI key

LIEOEYTUTSDYKB-BUHFOSPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermal inverse-type hetero-Diels-Alder reaction of O-silyl-protected lactal and bis(2,2,2-trichloroethyl) azodicarboxylate has been reported[1].

application

Bis(2,2,2-trichloroethyl) azodicarboxylate has been used:
  • in the asymmetric synthesis of substituted cycloalkyl[b]indoles[2]
  • as amination reagent during the aza-ene reaction of different alkenes to yield the corresponding allyl amines[3]
  • in para-directed amination of C2-alkyl substituted anisole[4]
  • in the synthesis of acid- and base-sensitive azo compounds and in Diels-Alder cycloadditions[5]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C Hillier et al.
The Journal of organic chemistry, 70(21), 8385-8394 (2005-10-08)
A general asymmetric synthesis of substituted cycloalkyl[b]indoles has been accomplished. The key features of this approach are (1) the utilization of a Japp-Klingemann condensation/Fischer cyclization to prepare cycloalkyl[b]indolones, (2) the asymmetric borane reduction of these heterocyclic ketones with (S)-OAB to
A Toepfer et al.
Carbohydrate research, 247, 159-164 (1993-09-02)
In a thermal inverse-type hetero-Diels-Alder reaction of O-silyl-protected lactal 1 and bis(2,2,2-trichloroethyl) azodicarboxylate (2), the dihydrooxadiazine derivative 3 was obtained in a very high yield; transesterification with benzyl alcohol furnished the corresponding derivative 4. Treatment of 3 with methanol in
Organic Syntheses, 61, 17-17 (1983)
Pompiliu S Aburel et al.
Organic & biomolecular chemistry, 3(12), 2344-2349 (2005-07-13)
Lewis acids such as Cu(OTf)(2), Zn(OTf)(2), Yb(OTf)(3) and Nd(OTf)(3) catalyze the aza-ene reaction of alkenes with azodicarboxylates, giving the allylic amination adducts. The use of bis(2,2,2-trichloroethyl)azodicarboxylate as the amination reagent and Cu(OTf)(2) and Yb(OTf)(3) as the catalysts gave the aza-ene
Para-directed amination of electron-rich arenes with bis (2, 2, 2-trichloroethyl) azodicarboxylate.
Leblanc Y and Boudreault N.
The Journal of Organic Chemistry, 60(13), 4268-4271 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service