Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

286753

Sigma-Aldrich

1-Naphthyl butyrate

99%

Synonym(s):

Butanoic acid 1-naphthyl ester, Butyric acid 1-naphthyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2C10H7
CAS Number:
Molecular Weight:
214.26
Beilstein/REAXYS Number:
2558646
EC Number:
MDL number:
UNSPSC Code:
12352103

description

esterase substrate

assay

99%

refractive index

n20/D 1.571 (lit.)

bp

125-130 °C/1 mmHg (lit.)

density

1.091 g/mL at 20 °C (lit.)

SMILES string

CCCC(=O)Oc1cccc2ccccc12

InChI

1S/C14H14O2/c1-2-6-14(15)16-13-10-5-8-11-7-3-4-9-12(11)13/h3-5,7-10H,2,6H2,1H3

InChI key

XIVJNRXPRQKFRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service