Skip to Content
MilliporeSigma
All Photos(1)

Documents

284017

Sigma-Aldrich

Maltoheptaose hydrate

≥80%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O36 · xH2O
CAS Number:
Molecular Weight:
1153.00 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥80%

optical activity

[α]25/D +178°, c = 1 in H2O

concentration

≤10% in water

SMILES string

[H]O[H].OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)C(O)O[C@@H]7CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H72O36.H2O/c43-1-8-15(50)16(51)24(59)37(67-8)74-31-10(3-45)69-39(26(61)18(31)53)76-33-12(5-47)71-41(28(63)20(33)55)78-35-14(7-49)72-42(29(64)22(35)57)77-34-13(6-48)70-40(27(62)21(34)56)75-32-11(4-46)68-38(25(60)19(32)54)73-30-9(2-44)66-36(65)23(58)17(30)52;/h8-65H,1-7H2;1H2/t8-,9-,10-,11-,12-,13-,14-,15-,16+,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36?,37-,38-,39-,40-,41-,42-;/m1./s1

InChI key

QBUKDCVGFUMISC-VPMWRFTJSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hao Xu et al.
Journal of pharmaceutical sciences, 108(1), 525-537 (2018-11-25)
This work reports the measurement of the partition rate of a model drug, ibuprofen (IBU), from aqueous solutions into octanol in the absence and presence of common pharmaceutical excipients including glucose, lactose, maltoheptaose, hydroxypropyl methylcellulose, polyvinylpyrrolidone, polyvinylpyrrolidone-vinyl acetate, hydroxypropyl methylcellulose AS
Weili Wang et al.
Journal of pharmaceutical sciences, 109(8), 2553-2566 (2020-05-31)
The partition rate of 2 model compounds, 2-naphthol (NAP) and ritonavir (RTV), across the water-octanol (W/O) interface was determined from aqueous solutions with and without the presence of a variety of common pharmaceutical excipients. The NAP molecule was present as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service