279013
4-Amino-2-chlorobenzonitrile
99%
Synonym(s):
3-Chloro-4-cyanoaniline
About This Item
Recommended Products
assay
99%
mp
116-118 °C (lit.)
functional group
chloro
nitrile
SMILES string
Nc1ccc(C#N)c(Cl)c1
InChI
1S/C7H5ClN2/c8-7-3-6(10)2-1-5(7)4-9/h1-3H,10H2
InChI key
ZFBKYGFPUCUYIF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service