Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278521

Sigma-Aldrich

Gadolinium(III) chloride hexahydrate

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3 · 6H2O
CAS Number:
Molecular Weight:
371.70
EC Number:
MDL number:
UNSPSC Code:
12352302

assay

99.9%

density

2.42 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

PNYPSKHTTCTAMD-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvio Aime et al.
Inorganic chemistry, 36(14), 2992-3000 (1997-07-02)
The synthesis of three triazamacrocycles containing the pyridine moiety and three acetate pendant arms (PCTA) is reported. The three systems differ due to the number of carbon atoms in the macrocyclic ring forming ligands PCTA-[12], -[13], and -[14], endowed with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service