Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

277738

Sigma-Aldrich

Benzophenone-(carbonyl-13C)

99 atom % 13C

Synonym(s):

Diphenylmethanone-13C

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$407.00

$407.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$407.00

About This Item

Linear Formula:
(C6H5)213CO
CAS Number:
Molecular Weight:
183.21
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$407.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

form

solid

bp

305 °C (lit.)

mp

47-51 °C (lit.)

mass shift

M+1

SMILES string

O=[13C](c1ccccc1)c2ccccc2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i13+1

InChI key

RWCCWEUUXYIKHB-KCKQSJSWSA-N

Gene Information

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

target_organs

Liver,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service