Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

273007

Sigma-Aldrich

γ-Methylene-γ-butyrolactone

99%

Synonym(s):

α′-Angelicalactone, 5-Methylenedihydro-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
Beilstein/REAXYS Number:
1560870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

99%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

80 °C/17 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

C=C1CCC(=O)O1

InChI

1S/C5H6O2/c1-4-2-3-5(6)7-4/h1-3H2

InChI key

SIFBVNDLLGPEKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The relative thermodynamic stabilities of γ-methylene-γ-butyrolactone were studied using chemical equilibration in cyclohexane and dipentyl ether solutions at several temperatures[1].

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relative thermodynamic stabilities of ?-methylene-?-butyrolactone and ?-angelicalactone.
Tetrahedron, 50(6), 1885-1888 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service