Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263370

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3CH2OH
CAS Number:
Molecular Weight:
244.13
Beilstein/REAXYS Number:
2055358
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

53-56 °C (lit.)

SMILES string

OCc1cc(cc(c1)C(F)(F)F)C(F)(F)F

InChI

1S/C9H6F6O/c10-8(11,12)6-1-5(4-16)2-7(3-6)9(13,14)15/h1-3,16H,4H2

InChI key

BJTWPJOGDWRYDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amide-based atropisomers in tachykinin NK1-receptor antagonists: synthesis and antagonistic activity of axially chiral N-benzylcarboxamide derivatives of 2,3,4,5-tetrahydro-6H-pyrido [2, 3-b][1,5] oxazocin-6-one.
Ishichi Y, et al.
Tetrahedron, 60(20), 4481-4490 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service