Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

258075

Sigma-Aldrich

2,2′,4,4′,4′′-Pentamethoxytriphenylmethanol

98%

Synonym(s):

2,2′,4,4′,4′′-Pentamethoxytrityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26O6
CAS Number:
Molecular Weight:
410.46
MDL number:
UNSPSC Code:
12352103

assay

98%

SMILES string

COc1ccc(cc1)C(O)(c2ccc(OC)cc2OC)c3ccc(OC)cc3OC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service