Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

253073

Sigma-Aldrich

D-(+)-Glucose

ACS reagent

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201

grade

ACS reagent

optical activity

[α]25/D +52.5 to +53.0°, c = 10 in water + trace NH4OH

impurities

≤0.002 meq/g
≤0.005% insolubles

ign. residue

≤0.02%

loss

≤0.2% loss on drying

mp

150-152 °C (lit.)

anion traces

SO42- and SO32-: ≤0.005%
chloride (Cl-): ≤0.01%

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Genin et al.
BMC cancer, 15, 577-577 (2015-08-09)
Tumor associated macrophages (TAMs) are present in high density in solid tumors. TAMs share many characteristics with alternatively activated macrophages, also called M2. They have been shown to favor tumor development and a role in chemoresistance has also been suggested.
Danielle Hagstrom et al.
Toxicological sciences : an official journal of the Society of Toxicology, 147(1), 270-285 (2015-06-28)
Traditional toxicology testing has relied on low-throughput, expensive mammalian studies; however, timely testing of the large number of environmental toxicants requires new in vitro and in vivo platforms for inexpensive medium- to high-throughput screening. Herein, we describe the suitability of the asexual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service