Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251909

Sigma-Aldrich

2-Ethylphenyl isocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4NCO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.5278 (lit.)

density

1.043 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1N=C=O

InChI

1S/C9H9NO/c1-2-8-5-3-4-6-9(8)10-7-11/h3-6H,2H2,1H3

InChI key

ZVFNUQWYLXXSJM-UHFFFAOYSA-N

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Fouad et al.
Journal of chromatography. A, 1515, 91-99 (2017-08-12)
Here we report the first encapsulation of three carbamylated amylose namely R-, S- and R/S-amylose 2,3(3,5-dimethylphenylcarbamate)-6-ethylphenylcarbamate in organic polymer monolith in situ capillary columns. The columns were investigated for the enantioselective nano-liquid chromatographic separation of a set of racemic pharmaceuticals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service