Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251623

Sigma-Aldrich

1,2-Dibromo-2-methylpropane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(Br)CH2Br
CAS Number:
Molecular Weight:
215.91
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.5 (vs air)

vapor pressure

15 mmHg ( 20 °C)

assay

98%

form

liquid

refractive index

n20/D 1.5093 (lit.)

bp

148-150 °C/763 mmHg (lit.)

density

1.75 g/mL at 25 °C (lit.)

SMILES string

CC(C)(Br)CBr

InChI

1S/C4H8Br2/c1-4(2,6)3-5/h3H2,1-2H3

InChI key

SDTXSEXYPROZSZ-UHFFFAOYSA-N

General description

Selective halogen exchange reactions of 1,2-dibromo-2-methylpropane with BiCl3 have been reported[1].

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BiX3 as an efficient and selective reagent for the halogen exchange reaction.
Boyer B, et al.
Tetrahedron, 55(7), 1971-1976 (1999)
Marzia Massignani et al.
PloS one, 5(5), e10459-e10459 (2010-05-11)
Microscopic techniques enable real-space imaging of complex biological events and processes. They have become an essential tool to confirm and complement hypotheses made by biomedical scientists and also allow the re-examination of existing models, hence influencing future investigations. Particularly imaging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service