Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250708

Sigma-Aldrich

2-(Trifluoromethyl)benzyl chloride

97%

Synonym(s):

α′-Chloro-α,α,α-trifluoro-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2Cl
CAS Number:
Molecular Weight:
194.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

liquid

refractive index

n20/D 1.469 (lit.)

density

1.337 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1CCl

InChI

1S/C8H6ClF3/c9-5-6-3-1-2-4-7(6)8(10,11)12/h1-4H,5H2

InChI key

BBXDMCQDLOCXRA-UHFFFAOYSA-N

Application

2-(Trifluoromethyl)benzyl chloride reacts with 2-benzo[b]thienyllithium to yield 6-fluorobenzo[b]naphtho[2,3-d]thiophene[1].

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An unconventional synthetic approach to fluoro heteroaromatic compounds by a novel transformation of an anionically activated trifluoromethyl group.
Kiselyov AS and Strekowski L.
Tetrahedron Letters, 35(41), 7597-7600 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service