Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248185

Sigma-Aldrich

2-Bromo-2-chloro-1,1,1-trifluoroethane

99%

Synonym(s):

Halothane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH(Cl)CF3
CAS Number:
Molecular Weight:
197.38
Beilstein/REAXYS Number:
1736947
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

4.5 psi ( 20 °C)

assay

99%

contains

0.01% thymol as stabilizer

refractive index

n20/D 1.369 (lit.)

bp

50.2 °C (lit.)

density

1.872 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(Cl)Br

InChI

1S/C2HBrClF3/c3-1(4)2(5,6)7/h1H

InChI key

BCQZXOMGPXTTIC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

2-Bromo-2-chloro-1,1,1-trifluoroethane is an inhalation anesthetic. In an effect believed to be independent of anesthesia, halothane inhibits the synthesis of 5-hydroxytryptamine in brain tissue, probably at the tryptophan hydroxylase step.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service