Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

247855

Sigma-Aldrich

Dimethyl methoxymalonate

98%

Synonym(s):

Trimethyl tartronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH(COOCH3)2
CAS Number:
Molecular Weight:
162.14
Beilstein/REAXYS Number:
1772841
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

refractive index

n20/D 1.423 (lit.)

bp

215 °C/745 mmHg (lit.)

density

1.172 g/mL at 25 °C (lit.)

SMILES string

COC(C(=O)OC)C(=O)OC

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

213.8 °F - closed cup

flash_point_c

101 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service