Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

243299

Sigma-Aldrich

1-Bromo-2-butanone

95%

Synonym(s):

Bromomethyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH2Br
CAS Number:
Molecular Weight:
151.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

liquid

contains

~5% calcium carbonate as stabilizer

refractive index

n20/D 1.465 (lit.)

bp

105 °C/150 mmHg (lit.)

density

1.479 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCC(=O)CBr

InChI

1S/C4H7BrO/c1-2-4(6)3-5/h2-3H2,1H3

InChI key

CCXQVBSQUQCEEO-UHFFFAOYSA-N

Application

1-Bromo-2-butanone (bromomethyl ethyl ketone) was used as a potential regulatory site-directed reagent for the residue C221 on pyruvate decarboxylase[1]. It was also used as a reagent for aromatic bromination with sodium hydride in DMSO[2] and in a microwave-assisted preparation of fused heterocycles[3].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 3889-3889 (2006)
Erin F DiMauro et al.
The Journal of organic chemistry, 71(10), 3959-3962 (2006-05-06)
The rapid and efficient synthesis of various disubstituted 5,6-fused heterocycles using a microwave-assisted one-pot cyclization-Suzuki coupling approach is described. This work highlights the tolerance of the boronic ester functional group to a variety of reaction conditions and the utility of
I Baburina et al.
Biochemistry, 37(5), 1245-1255 (1998-03-07)
The residue C221 on pyruvate decarboxylase (EC. 4.1.1.1) from Saccharomyces cerevisiae has been shown to be the site where the substrate activation cascade is triggered [Baburina et al. (1994) Biochemistry 33, 5630-5635] and is located on the beta domain [Arjunan

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service