239992
Dibromomethane
≥99%
Synonym(s):
Methylene bromide
About This Item
Recommended Products
vapor density
6.05 (vs air)
vapor pressure
34.9 mmHg ( 20 °C)
assay
≥99%
refractive index
n20/D 1.541 (lit.)
bp
96-98 °C (lit.)
mp
−52 °C (lit.)
solubility
water: 11.70 g/kg at 15 °C(lit.)
water: 11.93 g/kg at 30 °C(lit.)
acetone: miscible(lit.)
alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
density
2.477 g/mL at 25 °C (lit.)
SMILES string
BrCBr
InChI
1S/CH2Br2/c2-1-3/h1H2
InChI key
FJBFPHVGVWTDIP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 3
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service