Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

236721

Sigma-Aldrich

3-Methoxy-5-methylphenol

Synonym(s):

3-Hydroxy-5-methoxytoluene, 5-Methoxy-m-cresol, Orcinol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OCH3)OH
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12352100

mp

59-63 °C (lit.)

SMILES string

COc1cc(C)cc(O)c1

InChI

1S/C8H10O2/c1-6-3-7(9)5-8(4-6)10-2/h3-5,9H,1-2H3

InChI key

NOTCZLKDULMKBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service