Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235660

Sigma-Aldrich

Bis(dimethylamino)dimethylsilane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]2Si(CH3)2
CAS Number:
Molecular Weight:
146.31
Beilstein/REAXYS Number:
1736095
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

1 mmHg ( 20 °C)

assay

96%

refractive index

n20/D 1.417 (lit.)

bp

128-129 °C (lit.)

mp

−98 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CN(C)[Si](C)(C)N(C)C

InChI

1S/C6H18N2Si/c1-7(2)9(5,6)8(3)4/h1-6H3

Inchi Key

QULMGWCCKILBTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

19.4 °F - closed cup

flash_point_c

-7 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service