23420
5-Chloro-2-methoxyaniline
≥98.0% (HPLC), technical
Synonym(s):
2-Amino-4-chloroanisole, 5-Chloro-o-anisidine
About This Item
grade
technical
assay
≥98.0% (HPLC)
mp
81-83 °C (lit.)
81-83 °C
SMILES string
COc1ccc(Cl)cc1N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
276.8 °F - closed cup
flash_point_c
136 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service