Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

232610

Methyl acetopyruvate

97%

Synonym(s):

1-Methoxy-1,2,4-pentanetrione, Methyl β-acetylpyruvate, Methyl 2,4-dioxopentanoate, Methyl 2,4-dioxovalerate, Methyl 3-acetylpyruvate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

mp

60-65 °C

SMILES string

COC(=O)C(=O)CC(C)=O

InChI

1S/C6H8O4/c1-4(7)3-5(8)6(9)10-2/h3H2,1-2H3

InChI key

OMHOEQINEXASKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service