229016
Benzyldimethylstearylammonium chloride monohydrate
90%
About This Item
Recommended Products
assay
90%
mp
67-69 °C (lit.)
SMILES string
O.[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
InChI
1S/C27H50N.ClH.H2O/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-25-28(2,3)26-27-23-20-19-21-24-27;;/h19-21,23-24H,4-18,22,25-26H2,1-3H3;1H;1H2/q+1;;/p-1
InChI key
MEAWKEQKQGFJHS-UHFFFAOYSA-M
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service