Skip to Content
MilliporeSigma
All Photos(1)

Documents

228109

Sigma-Aldrich

Barium cyclohexanebutyrate

Synonym(s):

Barium 4-cyclohexylbutyrate, Bis(cyclohexanebutanoic acid)barium salt, Cyclohexanebutyric acid barium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H11(CH2)3CO2]2Ba
CAS Number:
Molecular Weight:
475.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

powder and chunks

mp

225 °C (dec.) (lit.)

SMILES string

O=C(CCCC1CCCCC1)O[Ba]OC(=O)CCCC2CCCCC2

InChI

1S/2C10H18O2.Ba/c2*11-10(12)8-4-7-9-5-2-1-3-6-9;/h2*9H,1-8H2,(H,11,12);/q;;+2/p-2

InChI key

NFAVTTVOXXCARJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Barium standard salt for organic systems. Lot specific assay on label.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Ougham et al.
Journal of bacteriology, 150(3), 1172-1182 (1982-06-01)
A strain of Arthrobacter was isolated by enrichment culture with cyclohexaneacetate as the sole source of carbon and grew with a doubling time of 4.2 h. In addition to growing with cyclohexaneacetate, the organism also grew with cyclohexanebutyrate at concentrations
Dean M Quesnel et al.
Chemosphere, 84(4), 504-511 (2011-04-05)
Naphthenic acids (NAs) are a major contributor to toxicity in tailings waste generated from bitumen production in the Athabasca Oil Sands region. While investigations have shown that bacteria can biodegrade NAs and reduce tailings toxicity, the potential of algae to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service