Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

228028

Sigma-Aldrich

1,8-Naphthosultam

97%

Synonym(s):

1,8-Naphthalenesultam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO2S
CAS Number:
Molecular Weight:
205.23
Beilstein/REAXYS Number:
83801
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

173-175 °C (lit.)

SMILES string

O=S1(=O)Nc2cccc3cccc1c23

InChI

1S/C10H7NO2S/c12-14(13)9-6-2-4-7-3-1-5-8(11-14)10(7)9/h1-6,11H

InChI key

HOBYPSNNFBRLIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,8-Naphthosultamyl group is a novel PBP2a (penicillin-binding protein 2a)-binding, anti-methicillin-resistant Staphylococcus aureus (MRSA) pharmacophore[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R R Wilkening et al.
Bioorganic & medicinal chemistry letters, 9(5), 673-678 (1999-04-14)
A series of 1beta-methyl carbapenems substituted at the 2-position with lipophilic, acyclic and cyclic (sulfonamido)methyl groups was prepared and evaluated for activity against resistant gram-positive bacteria. From these studies, the 1,8-naphthosultamyl group emerged as a novel, PBP2a-binding, anti-MRSA pharmacophore worthy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service