Skip to Content
MilliporeSigma
All Photos(1)

Documents

227242

Sigma-Aldrich

Cyclopropyl phenyl sulfide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5SC6H5
CAS Number:
Molecular Weight:
150.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

refractive index

n20/D 1.581 (lit.)

bp

62-63 °C/1 mmHg (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

C1CC1Sc2ccccc2

InChI

1S/C9H10S/c1-2-4-8(5-3-1)10-9-6-7-9/h1-5,9H,6-7H2

InChI key

YIBKCPJOFAUAKY-UHFFFAOYSA-N

Application

Cyclopropyl phenyl sulfide was used as a reagent in the conversion of a ketone into a spirocyclobutanone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 999-999 (2007)
Bumgardner CL, et al.
Tetrahedron Letters, 23(23), 2379-2382 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service