Skip to Content
MilliporeSigma
All Photos(1)

Documents

225622

Sigma-Aldrich

4-Amino-3,5,6-trichloropyridine-2-carboxylic acid

technical grade

Synonym(s):

4-Amino-3,5,6-trichloropicolinic acid, Picloram

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3N2O2
CAS Number:
Molecular Weight:
241.46
Beilstein/REAXYS Number:
479075
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

technical grade

mp

200 °C (dec.) (lit.)

SMILES string

Nc1c(Cl)c(Cl)nc(C(O)=O)c1Cl

InChI

1S/C6H3Cl3N2O2/c7-1-3(10)2(8)5(9)11-4(1)6(12)13/h(H2,10,11)(H,12,13)

InChI key

NQQVFXUMIDALNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Alayón-Luaces et al.
Plant science : an international journal of experimental plant biology, 185-186, 169-175 (2012-02-14)
The cell wall composition of apples callus cultures showed changes in the presence of 5 mg l(-1) of three different plant growth regulators (PGRs), namely picloram, abscisic acid and gibberellic acid. Although the structural functions of cell walls do not
Dean E Pearson et al.
Ecological applications : a publication of the Ecological Society of America, 18(6), 1489-1500 (2008-09-05)
Small-mammal seed predation is an important force structuring native-plant communities that may also influence exotic-plant invasions. In the intermountain West, deer mice (Peromyscus maniculatus) are prominent predators of native-plant seeds, but they avoid consuming seeds of certain widespread invasives like
Romina Sellaro et al.
Molecular plant, 5(3), 619-628 (2012-02-09)
We investigated the diurnal dependence of the hypocotyl-growth responses to shade under sunlight-night cycles in Arabidopsis thaliana. Afternoon shade events promoted hypocotyl growth, while morning shade was ineffective. The lhy-D, elf3, lux, pif4 pif5, toc1, and quadruple della mutants retained
Luz Irina A Calderón Villalobos et al.
Nature chemical biology, 8(5), 477-485 (2012-04-03)
The plant hormone auxin regulates virtually every aspect of plant growth and development. Auxin acts by binding the F-box protein transport inhibitor response 1 (TIR1) and promotes the degradation of the AUXIN/INDOLE-3-ACETIC ACID (Aux/IAA) transcriptional repressors. Here we show that
Ali Ozcan et al.
Journal of hazardous materials, 153(1-2), 718-727 (2007-10-16)
The degradation of the picloram, a widely used herbicide, has been undertaken by the electrochemical advanced oxidation process, namely electro-Fenton in aqueous solution. This process generates catalytically hydroxyl radicals that are strong oxidizing reagents for the oxidation of organic substances.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service