About This Item
Recommended Products
assay
98%
form
solid
mp
193-195 °C (lit.)
SMILES string
CC(=O)c1ccc(cc1)-c2ccc(cc2)C(C)=O
InChI
1S/C16H14O2/c1-11(17)13-3-7-15(8-4-13)16-9-5-14(6-10-16)12(2)18/h3-10H,1-2H3
Inchi Key
YSTSBXDVNKYPTR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service