Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219207

Sigma-Aldrich

N-Benzyl-tert-butylamine

97%

Synonym(s):

N-tert-Butylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHC(CH3)3
CAS Number:
Molecular Weight:
163.26
Beilstein/REAXYS Number:
507464
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.497 (lit.)

bp

80 °C/5 mmHg (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)NCc1ccccc1

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

194.0 °F

flash_point_c

90 °C

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ulgen et al.
Drug metabolism and drug interactions, 12(2), 131-143 (1995-01-01)
The metabolism of N-benzyl-tert-butylamine was studied in vitro using male hamster hepatic microsomal preparations. This substrate produced the corresponding nitrone, benzaldehyde and an uncharacterised metabolite. No metabolites were detected which corresponded to either authentic amide or oxaziridine. The results indicate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service